missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one, 97%

Catalog No. AAB2108614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2108614 25 g
AAB2108606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2108614 Supplier Thermo Scientific Chemicals Supplier No. B2108614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5381-99-7 | C7H4ClO3P | 202.53 g/mol

2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.

Solubility
Soluble in acetonitrile, dichloromethane, tetrahydrofuron and dimethylformamide.

Notes
Light and heat sensitive. Incompatible with water and strong oxidizing agents.

Chemical Identifiers

CAS 5381-99-7
Molecular Formula C7H4ClO3P
Molecular Weight (g/mol) 202.53
MDL Number MFCD00013353
InChI Key BVOITXUNGDUXRW-UHFFFAOYNA-N
Synonym 2-chloro-4h-1,3,2-benzodioxaphosphorin-4-one, 2-chloro-4h-benzo d 1,3,2 dioxaphosphinin-4-one, van boom's reagent, 2-chloro-4h-1,2,3-benzodioxaphosphorin-4-one, 4h-1,3,2-benzodioxaphosphorin-4-one, 2-chloro, salicylchlorophosphite, salicyl chlorophosphite, acmc-1an3b, 2-chloro-4h-1,3,2-benzodioxaphosphinin-4-one, salicylchlorophosphite; van boom's reagent
PubChem CID 237010
IUPAC Name 2-chloro-1,3,2-benzodioxaphosphinin-4-one
SMILES ClP1OC(=O)C2=CC=CC=C2O1

Specifications

Melting Point 36°C to 40°C
Boiling Point 127°C to 128°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Soluble in acetonitrile,dichloromethane,tetrahydrofuron and dimethylformamide.
Formula Weight 202.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only