missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)pyrimidine 98.0+%, TCI America™

Catalog No. C24671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24671G 1 g
C24675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24671G Supplier TCI America Supplier No. C24671G

Chemical Identifiers

CAS 33034-67-2
Molecular Formula C5H2ClF3N2
Molecular Weight (g/mol) 182.53
MDL Number MFCD00115111
InChI Key FZRBTBCCMVNZBD-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl pyrimidine, pyrimidine, 2-chloro-4-trifluoromethyl, 4-trifluoromethyl-2-chloropyrimidine, 2-chloro-4-trifluoromethyl-pyrimidine, pubchem6907, acmc-209hxy, ksc222e4j, abbypharma ap-10-1130
PubChem CID 2773912
IUPAC Name 2-chloro-4-(trifluoromethyl)pyrimidine
SMILES C1=CN=C(N=C1C(F)(F)F)Cl

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
UN Number 1993
Formula Weight 182.53
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)pyrimidine