missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)pyrimidine, 97%

Catalog No. AC397850010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397850010 1 g Glass bottle
AC397850050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397850010 Supplier Thermo Scientific Chemicals Supplier No. 397850010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 33034-67-2 | C5H2ClF3N2 | 182.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33034-67-2
Molecular Formula C5H2ClF3N2
Molecular Weight (g/mol) 182.53
MDL Number MFCD00115111
InChI Key FZRBTBCCMVNZBD-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl pyrimidine, pyrimidine, 2-chloro-4-trifluoromethyl, 4-trifluoromethyl-2-chloropyrimidine, 2-chloro-4-trifluoromethyl-pyrimidine, pubchem6907, acmc-209hxy, ksc222e4j, abbypharma ap-10-1130
PubChem CID 2773912
IUPAC Name 2-chloro-4-(trifluoromethyl)pyrimidine
SMILES C1=CN=C(N=C1C(F)(F)F)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5130g/mL
Boiling Point 60°C (10.0 mmHg)
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.513
Formula Weight 182.53
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-chloro-4-(trifluoromethyl)pyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only