missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-(trifluoromethyl)pyridine 98.0+%, TCI America™
$111.66 - $392.74
Chemical Identifiers
| CAS | 81565-18-6 |
|---|---|
| Molecular Formula | C6H3ClF3N |
| Molecular Weight (g/mol) | 181.54 |
| MDL Number | MFCD00042224 |
| InChI Key | GBNPVXZNWBWNEN-UHFFFAOYSA-N |
| Synonym | 2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine |
| PubChem CID | 144958 |
| IUPAC Name | 2-chloro-4-(trifluoromethyl)pyridine |
| SMILES | FC(F)(F)C1=CC=NC(Cl)=C1 |
Chemical Identifiers
| 81565-18-6 | |
| 181.54 | |
| GBNPVXZNWBWNEN-UHFFFAOYSA-N | |
| 144958 | |
| FC(F)(F)C1=CC=NC(Cl)=C1 |
| C6H3ClF3N | |
| MFCD00042224 | |
| 2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine | |
| 2-chloro-4-(trifluoromethyl)pyridine |
Specifications
| 81565-18-6 | |
| 147°C | |
| MFCD00042224 | |
| 1992 | |
| GBNPVXZNWBWNEN-UHFFFAOYSA-N | |
| 2-chloro-4-(trifluoromethyl)pyridine | |
| 144958 | |
| ≥98.0% (GC) | |
| 2-Chloro-4-(trifluoromethyl)pyridine |
| Yellow | |
| C6H3ClF3N | |
| 1 g | |
| 2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine | |
| FC(F)(F)C1=CC=NC(Cl)=C1 | |
| 181.54 | |
| 181.54 | |
| Liquid |
Safety and Handling
TSCA : No