missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

Catalog No. AAH2731203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2731203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2731203 Supplier Thermo Scientific Chemicals Supplier No. H2731203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 590371-81-6
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.55
MDL Number MFCD08458125
InChI Key FZIJHEOXLKCIRZ-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl nicotinic acid, 2-chloro-4-trifluoromethyl pyridine-3-carboxylic acid, 3-carboxy-2-chloro-4-trifluoromethyl pyridine, acmc-1az7h, ksc269c2b, 3-pyridinecarboxylicacid, 2-chloro-4-trifluoromethyl, 3-pyridinecarboxylic acid, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoromethyl-3-pyridinecarboxylic acid
PubChem CID 11770138
IUPAC Name 2-chloro-4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=C(C=CN=C1Cl)C(F)(F)F

Specifications

Melting Point 158°C to 159°C
Quantity 1 g
Formula Weight 225.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)nicotinic acid
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures