missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitroaniline, 98+%

Catalog No. aaa165860b
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA165860B 1000 g
AAA1658630 250 g
AAA165860I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA165860B Supplier Thermo Scientific Chemicals Supplier No. A165860B
Only null left

CAS: 121-87-9 | C6H5ClN2O2 | 172.568 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-87-9
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007665
InChI Key LOCWBQIWHWIRGN-UHFFFAOYSA-N
Synonym ocpna, 2-chloro-4-nitrobenzenamine, o-chloro-p-nitroaniline, 4-nitro-2-chloroaniline, benzenamine, 2-chloro-4-nitro, 1-amino-2-chloro-4-nitrobenzene, aniline, 2-chloro-4-nitro, unii-xsa3zx337b, 2-chloro-4-nitro-phenylamine, ortho-chloro-para-nitroaniline
PubChem CID 8492
IUPAC Name 2-chloro-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)N

Specifications

Melting Point 106°C to 109°C
Boiling Point 320°C (decomposition)
Flash Point 193°C (379°F)
Odor Weak, Characteristic
Quantity 1000 g
UN Number UN2237
Beilstein 638657
Formula Weight 172.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-4-nitroaniline
Hazard Category H302-H500
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROANILINES
EINECSNumber 204-502-2
RTECSNumber BX1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only