missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-methylpyrimidine 98.0+%, TCI America™
$87.60 - $307.57
Chemical Identifiers
| CAS | 13036-57-2 |
|---|---|
| Molecular Formula | C5H5ClN2 |
| Molecular Weight (g/mol) | 128.559 |
| MDL Number | MFCD00054434 |
| InChI Key | BHAKRVSCGILCEW-UHFFFAOYSA-N |
| Synonym | 2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride |
| PubChem CID | 11629607 |
| IUPAC Name | 2-chloro-4-methylpyrimidine |
| SMILES | CC1=NC(=NC=C1)Cl |
Chemical Identifiers
| 13036-57-2 | |
| 128.559 | |
| BHAKRVSCGILCEW-UHFFFAOYSA-N | |
| 11629607 | |
| CC1=NC(=NC=C1)Cl |
| C5H5ClN2 | |
| MFCD00054434 | |
| 2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride | |
| 2-chloro-4-methylpyrimidine |
Specifications
| 13036-57-2 | |
| White-Yellow | |
| C5H5ClN2 | |
| 5 g | |
| BHAKRVSCGILCEW-UHFFFAOYSA-N | |
| 2-chloro-4-methylpyrimidine | |
| 11629607 | |
| ≥98.0% (GC) | |
| 2-Chloro-4-methylpyrimidine |
| 49°C | |
| 94°C | |
| MFCD00054434 | |
| 2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride | |
| CC1=NC(=NC=C1)Cl | |
| 128.559 | |
| 128.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No