missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylpyrimidine, 99%

Catalog No. AAH3190303
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3190303 1 g
AAH3190309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3190303 Supplier Thermo Scientific Chemicals Supplier No. H3190303

CAS: 13036-57-2 | C5H5ClN2 | 128.559 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13036-57-2
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00054434
InChI Key BHAKRVSCGILCEW-UHFFFAOYSA-N
Synonym 2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride
PubChem CID 11629607
IUPAC Name 2-chloro-4-methylpyrimidine
SMILES CC1=NC(=NC=C1)Cl

Specifications

Melting Point 45°C to 48°C
Quantity 1 g
Formula Weight 128.56
Percent Purity 99%
Chemical Name or Material 2-Chloro-4-methylpyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only