missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog No. p-4812774
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453340100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453340100 Supplier Thermo Scientific Chemicals Supplier No. 453340100
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13036-57-2
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
InChI Key BHAKRVSCGILCEW-UHFFFAOYSA-N
Synonym 2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride
PubChem CID 11629607
IUPAC Name 2-chloro-4-methylpyrimidine
SMILES CC1=NC(=NC=C1)Cl

Specifications

Melting Point 51°C to 56°C
Boiling Point 93°C to 95°C (17.0 mmHg)
Assay Percent Range 97.5% min. (GC)
Quantity 10 g
Formula Weight 128.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methylpyrimidine
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only