missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-methylpyridine 98.0+%, TCI America™
$50.33 - $139.81
Chemical Identifiers
| CAS | 3678-62-4 |
|---|---|
| Molecular Formula | C6H6ClN |
| Molecular Weight (g/mol) | 127.571 |
| InChI Key | MZVSTDHRRYQFGI-UHFFFAOYSA-N |
| Synonym | 2-chloro-4-picoline, pyridine, 2-chloro-4-methyl, 2-chloro-4-methyl-pyridine, 2-chloro-4-methyl pyridine, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, zlchem 438, pubchem1192, acmc-209iop |
| PubChem CID | 77248 |
| IUPAC Name | 2-chloro-4-methylpyridine |
| SMILES | CC1=CC(=NC=C1)Cl |
Chemical Identifiers
| 3678-62-4 | |
| 127.571 | |
| 2-chloro-4-picoline, pyridine, 2-chloro-4-methyl, 2-chloro-4-methyl-pyridine, 2-chloro-4-methyl pyridine, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, zlchem 438, pubchem1192, acmc-209iop | |
| 2-chloro-4-methylpyridine |
| C6H6ClN | |
| MZVSTDHRRYQFGI-UHFFFAOYSA-N | |
| 77248 | |
| CC1=CC(=NC=C1)Cl |
Specifications
| 3678-62-4 | |
| 99°C | |
| 5 g | |
| MZVSTDHRRYQFGI-UHFFFAOYSA-N | |
| 2-chloro-4-methylpyridine | |
| 77248 | |
| ≥98.0% (GC) | |
| 2-Chloro-4-methylpyridine |
| Yellow | |
| C6H6ClN | |
| 2-chloro-4-picoline, pyridine, 2-chloro-4-methyl, 2-chloro-4-methyl-pyridine, 2-chloro-4-methyl pyridine, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, zlchem 438, pubchem1192, acmc-209iop | |
| CC1=CC(=NC=C1)Cl | |
| 127.571 | |
| 127.57 | |
| Liquid |
Safety and Handling
TSCA : No