missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. C20121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20121G 1 g
C20125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20121G Supplier TCI America Supplier No. C20121G
Only null left

Chemical Identifiers

CAS 23056-39-5
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
InChI Key JHARVUVBTAAPLA-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-4-picoline, pyridine, 2-chloro-4-methyl-3-nitro, 2-chloro-3-nitro-4-methylpyridine, 2-chloro-4-methyl-3-nitro-pyridine, pubchem6148, acmc-1cui4, 2-chloro-3-nitro4-picoline, ksc496c3n, 2-chlor-4-methyl-3-nitropyridin
PubChem CID 345363
IUPAC Name 2-chloro-4-methyl-3-nitropyridine
SMILES CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 53°C
Color Yellow
Quantity 1 g
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methyl-3-nitropyridine