missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxypyrimidine, 98%

Catalog No. AC426820010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426820010 1 g Glass bottle
AC426820050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426820010 Supplier Thermo Scientific Chemicals Supplier No. 426820010

CAS: 22536-63-6 | C5H5ClN2O | 144.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22536-63-6
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.56
MDL Number MFCD00194055
InChI Key BDXYNMVQMBCTDB-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxy-pyrimidine, pyrimidine, 2-chloro-4-methoxy, acmc-20a0n4, 2-chloro4-methoxypyrimidine, 4-methoxy-2-chloropyrimidine, ksc494k1r, 2-chloro-4-methoxypyrimidine, 2-chloro-4-methyloxy pyrimidine, 2-chloro-4-methoxy-1,2-diazine
PubChem CID 588935
IUPAC Name 2-chloro-4-methoxypyrimidine
SMILES COC1=NC(=NC=C1)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54°C to 57°C
Color White to Yellow
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 144.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methoxypyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only