missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxypyridine 98.0+%, TCI America™

Catalog No. C30245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30245G 5 g
C30241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30245G Supplier TCI America Supplier No. C30245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17228-69-2
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD02093951
InChI Key PMTPFBWHUOWTNN-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxy-pyridine, pyridine, 2-chloro-4-methoxy, zlchem 749, pubchem6204, 4-methoxy-2-chloropyridine, 2-chloro-4-methoxylpyridine, 2-chloro-4-methoxy pyridine, ksc494i4l, 2-chloro-4-methoxypyridine
PubChem CID 1991128
IUPAC Name 2-chloro-4-methoxypyridine
SMILES COC1=CC(=NC=C1)Cl

Specifications

Color Yellow
Boiling Point 230°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-methoxypyridine