missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxypyridine, 99%, Thermo Scientific™

$263.03 - $263.03

Chemical Identifiers

CAS 17228-69-2
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD02093951
InChI Key PMTPFBWHUOWTNN-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxy-pyridine, pyridine, 2-chloro-4-methoxy, zlchem 749, pubchem6204, 4-methoxy-2-chloropyridine, 2-chloro-4-methoxylpyridine, 2-chloro-4-methoxy pyridine, ksc494i4l, 2-chloro-4-methoxypyridine
PubChem CID 1991128
IUPAC Name 2-chloro-4-methoxypyridine
SMILES COC1=CC(=NC=C1)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1769703
View Documents
Thermo Scientific Chemicals
L1769703
1 g
Each for $263.03
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an active pharmaceutical ingredient.

Solubility
Soluble in alcohol.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
Specifications

Chemical Identifiers

17228-69-2
143.57
PMTPFBWHUOWTNN-UHFFFAOYSA-N
1991128
COC1=CC(=NC=C1)Cl
C6H6ClNO
MFCD02093951
2-chloro-4-methoxy-pyridine, pyridine, 2-chloro-4-methoxy, zlchem 749, pubchem6204, 4-methoxy-2-chloropyridine, 2-chloro-4-methoxylpyridine, 2-chloro-4-methoxy pyridine, ksc494i4l, 2-chloro-4-methoxypyridine
2-chloro-4-methoxypyridine

Specifications

17228-69-2
110°C to 115°C (10 mmHg)
C6H6ClNO
MFCD02093951
1423379
Soluble in alcohol.
COC1=CC(=NC=C1)Cl
143.57
143.57
2-Chloro-4-methoxypyridine
1.258
>110°C (230°F)
1.541
1 g
2-chloro-4-methoxy-pyridine, pyridine, 2-chloro-4-methoxy, zlchem 749, pubchem6204, 4-methoxy-2-chloropyridine, 2-chloro-4-methoxylpyridine, 2-chloro-4-methoxy pyridine, ksc494i4l, 2-chloro-4-methoxypyridine
PMTPFBWHUOWTNN-UHFFFAOYSA-N
2-chloro-4-methoxypyridine
1991128
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'