missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxypyridine, 99%, Thermo Scientific™

Catalog No. AAL1769703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1769703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1769703 Supplier Thermo Scientific Chemicals Supplier No. L1769703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an active pharmaceutical ingredient.

Solubility
Soluble in alcohol.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.

Chemical Identifiers

CAS 17228-69-2
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD02093951
InChI Key PMTPFBWHUOWTNN-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxy-pyridine, pyridine, 2-chloro-4-methoxy, zlchem 749, pubchem6204, 4-methoxy-2-chloropyridine, 2-chloro-4-methoxylpyridine, 2-chloro-4-methoxy pyridine, ksc494i4l, 2-chloro-4-methoxypyridine
PubChem CID 1991128
IUPAC Name 2-chloro-4-methoxypyridine
SMILES COC1=CC(=NC=C1)Cl

Specifications

Density 1.258
Boiling Point 110°C to 115°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.541
Quantity 1 g
Beilstein 1423379
Solubility Information Soluble in alcohol.
Formula Weight 143.57
Percent Purity 99%
Chemical Name or Material 2-Chloro-4-methoxypyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures