missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-4-methoxypyridine, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Boiling Point | 229°C to 230°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Quantity | 1g |
| Formula Weight | 143.57 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Chloro-4-methoxypyridine |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor |
RUO – Research Use Only