missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193785
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23861G Supplier TCI America Supplier No. C23861G
Only null left

Chemical Identifiers

CAS 219735-99-6
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD03411935
InChI Key LOCGPWGCRVKCFN-UHFFFAOYSA-N
PubChem CID 2773330
IUPAC Name (2-chloro-4-methoxyphenyl)boronic acid
SMILES COC1=CC=C(B(O)O)C(Cl)=C1

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 1 g
Synonym 2-chloro-4-methoxyphenyl boronic acid, 2-chloro-4-methoxybenzeneboronic acid, 4-borono-3-chloroanisole, 2-chloro-4-methoxyphenyl-boronic acid, boronic acid, 2-chloro-4-methoxyphenyl, 2-chloro-4-methoxyphenylboronicacid, pubchem1781, 2-chloro-4-methoxy-phenyl boronic acid, acmc-209fq2, ksc201q2h
Formula Weight 186.40
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride)