missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxybenzeneboronic acid, 95%, Thermo Scientific Chemicals

Catalog No. AAH2691706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2691706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2691706 Supplier Thermo Scientific Chemicals Supplier No. H2691706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 219735-99-6 | Molecular Formula: C7H8BClO3 | Molecular Weight: 186.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 219735-99-6
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD03411935
InChI Key LOCGPWGCRVKCFN-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxyphenyl boronic acid, 2-chloro-4-methoxybenzeneboronic acid, 4-borono-3-chloroanisole, 2-chloro-4-methoxyphenyl-boronic acid, boronic acid, 2-chloro-4-methoxyphenyl, 2-chloro-4-methoxyphenylboronicacid, pubchem1781, 2-chloro-4-methoxy-phenyl boronic acid, acmc-209fq2, ksc201q2h
PubChem CID 2773330
IUPAC Name (2-chloro-4-methoxyphenyl)boronic acid
SMILES COC1=CC=C(B(O)O)C(Cl)=C1

Specifications

Melting Point ∼202°C
Quantity 5 g
Formula Weight 186.4
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only