missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-iodo-5-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. C29995G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29995G 5 g
C29991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29995G Supplier TCI America Supplier No. C29995G

Chemical Identifiers

CAS 505084-55-9
Molecular Formula C6H2ClF3IN
Molecular Weight (g/mol) 307.439
MDL Number MFCD01862099
InChI Key XEEQBBXEZSNIQV-UHFFFAOYSA-N
PubChem CID 2773791
IUPAC Name 2-chloro-4-iodo-5-(trifluoromethyl)pyridine
SMILES C1=C(C(=CN=C1Cl)C(F)(F)F)I

Specifications

Melting Point 128°C
Color Yellow
Quantity 5 g
Synonym 2-chloro-4-iodo-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl-4-iodopyridine, 2-chloro-4-iodo-5-trifluoromethyl-pyridine, pyridine, 2-chloro-4-iodo-5-trifluoromethyl, acmc-1avx5, ksc496i6b, pyridine,2-chloro-4-iodo-5-trifluoromethyl
Formula Weight 307.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine