missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-hydroxypyridine 97.0+%, TCI America™

Catalog No. C25165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25165G 5 g
C25161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25165G Supplier TCI America Supplier No. C25165G

Chemical Identifiers

CAS 17368-12-6
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD01646108
InChI Key VBEHFOMFHUQAOW-UHFFFAOYSA-N
Synonym 2-chloro-4-hydroxypyridine, 2-chloropyridin-4-ol, 2-chloro-4-pyridinol, 4-pyridinol, 2-chloro, 2-chlorpyridin-4-ol, 4 1h-pyridinone, 2-chloro, pubchem2578, acmc-1btyf, 4-hydroxy-2-chloropyridine
PubChem CID 87010
IUPAC Name 2-chloro-1H-pyridin-4-one
SMILES C1=CNC(=CC1=O)Cl

Specifications

Melting Point 169°C
Quantity 5 g
Formula Weight 129.54
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-hydroxypyridine