missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-hydroxypyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH2754903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2754903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2754903 Supplier Thermo Scientific Chemicals Supplier No. H2754903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17368-12-6
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD01646108
InChI Key VBEHFOMFHUQAOW-UHFFFAOYSA-N
Synonym 2-chloro-4-hydroxypyridine, 2-chloropyridin-4-ol, 2-chloro-4-pyridinol, 4-pyridinol, 2-chloro, 2-chlorpyridin-4-ol, 4 1h-pyridinone, 2-chloro, pubchem2578, acmc-1btyf, 4-hydroxy-2-chloropyridine
PubChem CID 87010
IUPAC Name 2-chloro-1H-pyridin-4-one
SMILES C1=CNC(=CC1=O)Cl

Specifications

Melting Point 169°C to 172°C
Quantity 1 g
Formula Weight 129.55
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-399-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only