missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-hydroxybenzoic acid hydrate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 304200050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC304200050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

56363-84-9
172.57
WIPYZRZPNMUSER-UHFFFAOYSA-N
91821
C1=CC(=C(C=C1O)Cl)C(=O)O
C7H5ClO3·xH2O
MFCD00798116
benzoic acid, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzoic acid, 2-chloro-4-hyroxybenzoic acid, pubchem4561, acmc-1b14n, ksc497o6l, 4-hydroxy-2-chlorobenzoic acid, wipyzrzpnmuser-uhfffaoysa
2-chloro-4-hydroxybenzoic acid

Specifications

2-chloro-4-hydroxybenzoic acid hydrate
97.5
Authentic
Glass bottle
5g
benzoic acid, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzoic acid, 2-chloro-4-hyroxybenzoic acid, pubchem4561, acmc-1b14n, ksc497o6l, 4-hydroxy-2-chlorobenzoic acid, wipyzrzpnmuser-uhfffaoysa
C1=CC(=C(C=C1O)Cl)C(=O)O
172.57
172.57
56363-84-9
100.0
98%
C7H5ClO3·xH2O
MFCD00798116
WIPYZRZPNMUSER-UHFFFAOYSA-N
2-chloro-4-hydroxybenzoic acid
91821
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 260-132-1

SDS
Documents

Documents