missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-hydroxybenzoic acid hydrate, 98%, Thermo Scientific™

Catalog No. AC304200010
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304200010 1g Glass bottle
AC304200050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC304200010 Supplier Thermo Scientific Chemicals Supplier No. 304200010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 56363-84-9
Molecular Formula C7H5ClO3·xH2O
Molecular Weight (g/mol) 172.57
MDL Number MFCD00798116
InChI Key WIPYZRZPNMUSER-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzoic acid, 2-chloro-4-hyroxybenzoic acid, pubchem4561, acmc-1b14n, ksc497o6l, 4-hydroxy-2-chlorobenzoic acid, wipyzrzpnmuser-uhfffaoysa
PubChem CID 91821
IUPAC Name 2-chloro-4-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)Cl)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1g
Formula Weight 172.57
Percent Purity 98%
Chemical Name or Material 2-chloro-4-hydroxybenzoic acid hydrate
EINECSNumber 260-132-1

RUO – Research Use Only