missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-hydroxybenzaldehyde, 97%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 398030010

Catalog No. AC398030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Chloro-4-hydroxybenzaldehyde
96% min. (GC)
C7H5ClO2
1g
ZMOMCILMBYEGLD-UHFFFAOYSA-N
2-chloro-4-hydroxybenzaldehyde
185363
97%
56962-11-9
Glass Bottle
ClC6H3(OH)CHO
benzaldehyde, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzaldehyde, benzaldehyde, chloro-4-hydroxy, 2-chloro-4-hydroxybenzaldehyde, pubchem7366, acmc-209lv6, ksc497o6r, 2-chlor-4-hydroxy-benzaldehyd, 2-chloranyl-4-oxidanyl-benzaldehyde
C1=CC(=C(C=C1O)Cl)C=O
156.57
156.57

Chemical Identifiers

56962-11-9
156.57
benzaldehyde, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzaldehyde, benzaldehyde, chloro-4-hydroxy, 2-chloro-4-hydroxybenzaldehyde, pubchem7366, acmc-209lv6, ksc497o6r, 2-chlor-4-hydroxy-benzaldehyd, 2-chloranyl-4-oxidanyl-benzaldehyde
2-chloro-4-hydroxybenzaldehyde
C7H5ClO2
ZMOMCILMBYEGLD-UHFFFAOYSA-N
185363
C1=CC(=C(C=C1O)Cl)C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only