missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. C21295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21295G 5 g
C212925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21295G Supplier TCI America Supplier No. C21295G

Chemical Identifiers

CAS 60702-69-4
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.56
MDL Number MFCD00042523
InChI Key PGKPNNMOFHNZJX-UHFFFAOYSA-N
Synonym 4-fluoro-2-chlorobenzonitrile, benzonitrile, 2-chloro-4-fluoro, 2-chloro-4-fluoro benzonitrile, 2-chloro-4-fluoro-benzonitrile, 2-chloro-4-fluorobenzenecarbonitrile, ncr bg df, pubchem3422, acmc-209mll, 2-chloro-4fluorobenzonitrile, 2-chloro4-fluorobenzonitrile
PubChem CID 109000
IUPAC Name 2-chloro-4-fluorobenzonitrile
SMILES FC1=CC=C(C#N)C(Cl)=C1

Specifications

Melting Point 65°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Formula Weight 155.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chloro-4-fluorobenzonitrile