missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzonitrile, 98%

Catalog No. p-7022614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1547806 5 g
AAA1547814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1547806 Supplier Thermo Scientific Chemicals Supplier No. A1547806
Only null left

CAS: 60702-69-4 | C7H3ClFN | 155.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60702-69-4
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.56
MDL Number MFCD00042523
InChI Key PGKPNNMOFHNZJX-UHFFFAOYSA-N
Synonym 4-fluoro-2-chlorobenzonitrile, benzonitrile, 2-chloro-4-fluoro, 2-chloro-4-fluoro benzonitrile, 2-chloro-4-fluoro-benzonitrile, 2-chloro-4-fluorobenzenecarbonitrile, ncr bg df, pubchem3422, acmc-209mll, 2-chloro-4fluorobenzonitrile, 2-chloro4-fluorobenzonitrile
PubChem CID 109000
IUPAC Name 2-chloro-4-fluorobenzonitrile
SMILES FC1=CC=C(C#N)C(Cl)=C1

Specifications

Melting Point 62°C to 66°C
Quantity 5 g
UN Number UN3439
Beilstein 6323034
Formula Weight 155.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 262-384-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only