missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-fluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 85958-57-2
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD00052957
InChI Key FCFPJKHVCHKCMP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzene-1-sulfonyl chloride, 2-chloro-4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-4-fluoro, benzenesulfonyl chloride, 2-chloro-4-fluoro-9ci, pubchem5092, acmc-209wex, buttpark 9457-18, 2-chloro-4-fluorobenzene-1-sulfonylchloride, ksc447s0p, chloro 2-chloro-4-fluorophenyl sulfone
PubChem CID 522846
IUPAC Name 2-chloro-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)Cl)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
TL00318DA
SDS View Documents
Thermo Scientific Chemicals
TL00318DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

85958-57-2
229.046
FCFPJKHVCHKCMP-UHFFFAOYSA-N
522846
C1=CC(=C(C=C1F)Cl)S(=O)(=O)Cl
C6H3Cl2FO2S
MFCD00052957
2-chloro-4-fluorobenzene-1-sulfonyl chloride, 2-chloro-4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-4-fluoro, benzenesulfonyl chloride, 2-chloro-4-fluoro-9ci, pubchem5092, acmc-209wex, buttpark 9457-18, 2-chloro-4-fluorobenzene-1-sulfonylchloride, ksc447s0p, chloro 2-chloro-4-fluorophenyl sulfone
2-chloro-4-fluorobenzenesulfonyl chloride

Specifications

85958-57-2
97%
C6H3Cl2FO2S
MFCD00052957
2-chloro-4-fluorobenzene-1-sulfonyl chloride, 2-chloro-4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-4-fluoro, benzenesulfonyl chloride, 2-chloro-4-fluoro-9ci, pubchem5092, acmc-209wex, buttpark 9457-18, 2-chloro-4-fluorobenzene-1-sulfonylchloride, ksc447s0p, chloro 2-chloro-4-fluorophenyl sulfone
C1=CC(=C(C=C1F)Cl)S(=O)(=O)Cl
229.046
229.06
2-Chloro-4-fluorobenzene-1-sulfonyl chloride
97.0
Amber Glass Bottle
Cl(F)C6H3(SO2Cl)
1g
FCFPJKHVCHKCMP-UHFFFAOYSA-N
2-chloro-4-fluorobenzenesulfonyl chloride
522846
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'