missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4'-fluoroacetophenone 98.0+%, TCI America™
$37.29 - $123.03
Chemical Identifiers
| CAS | 456-04-2 |
|---|---|
| Molecular Formula | C8H6ClFO |
| Molecular Weight (g/mol) | 172.583 |
| InChI Key | UJZWJOQRSMOFMA-UHFFFAOYSA-N |
| Synonym | 2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone |
| PubChem CID | 120248 |
| IUPAC Name | 2-chloro-1-(4-fluorophenyl)ethanone |
| SMILES | C1=CC(=CC=C1C(=O)CCl)F |
Chemical Identifiers
| 456-04-2 | |
| 172.583 | |
| 2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone | |
| 2-chloro-1-(4-fluorophenyl)ethanone |
| C8H6ClFO | |
| UJZWJOQRSMOFMA-UHFFFAOYSA-N | |
| 120248 | |
| C1=CC(=CC=C1C(=O)CCl)F |
Specifications
| 456-04-2 | |
| White-Yellow | |
| C8H6ClFO | |
| 2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone | |
| C1=CC(=CC=C1C(=O)CCl)F | |
| 172.583 | |
| 172.58 | |
| Crystalline Powder |
| 50°C | |
| 247°C | |
| 5 g | |
| UJZWJOQRSMOFMA-UHFFFAOYSA-N | |
| 2-chloro-1-(4-fluorophenyl)ethanone | |
| 120248 | |
| ≥98.0% (GC) | |
| 2-Chloro-4′-fluoroacetophenone |
Safety and Handling
RTECSNumber : AM6550000
TSCA : No