missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4'-fluoroacetophenone 98.0+%, TCI America™

Catalog No. C19775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19775G 5 g
C197725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19775G Supplier TCI America Supplier No. C19775G

Chemical Identifiers

CAS 456-04-2
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
InChI Key UJZWJOQRSMOFMA-UHFFFAOYSA-N
Synonym 2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone
PubChem CID 120248
IUPAC Name 2-chloro-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CCl)F

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 247°C
Quantity 5 g
Formula Weight 172.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4′-fluoroacetophenone
RTECSNumber AM6550000
TSCA No