missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloro-4'-fluoroacetanilide 98.0+%, TCI America™

Catalog No. C131125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C131125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C131125G Supplier TCI America Supplier No. C131125G

Chemical Identifiers

CAS 399-35-9
Molecular Formula C8H7ClFNO
Molecular Weight (g/mol) 187.598
MDL Number MFCD00042594
InChI Key ZULZFLOGABTQFR-UHFFFAOYSA-N
Synonym 2'-chloro-4'-fluoroacetanilide, n-2-chloro-4-fluorophenyl acetamide, 2-chloro-4-fluoroacetanilide, acetamide, n-2-chloro-4-fluorophenyl, zulzflogabtqfr-uhfffaoysa, acetamide,n-2-chloro-4-fluorophenyl, n-2-chloro-4-fluorophenyl acetamide #, n-2-chloranyl-4-fluoranyl-phenyl ethanamide
PubChem CID 589419
IUPAC Name N-(2-chloro-4-fluorophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)F)Cl

Specifications

Melting Point 118°C
Quantity 25 g
Formula Weight 187.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Chloro-4′-fluoroacetanilide