missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-3-methylbenzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 173315-54-3
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.58
MDL Number MFCD11846444
InChI Key DBMUFFHLBXKTDM-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-fluoro-3-methyl, 2-chloro-4-fluoro-3-methyl-benzoic acid, 2-chloro-3-methyl-4-fluoro-benzoic acid, 2-chloro-4-fluoro-3-methyl benzoic acid
PubChem CID 10845315
IUPAC Name 2-chloro-4-fluoro-3-methylbenzoic acid
SMILES CC1=C(F)C=CC(C(O)=O)=C1Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5187903
View Documents
Thermo Scientific Chemicals
H5187903
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the synthesis and Antibacterial Evaluation of 6-Amino-8-methylquinolones.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

173315-54-3
188.58
DBMUFFHLBXKTDM-UHFFFAOYSA-N
10845315
CC1=C(F)C=CC(C(O)=O)=C1Cl
C8H6ClFO2
MFCD11846444
benzoic acid, 2-chloro-4-fluoro-3-methyl, 2-chloro-4-fluoro-3-methyl-benzoic acid, 2-chloro-3-methyl-4-fluoro-benzoic acid, 2-chloro-4-fluoro-3-methyl benzoic acid
2-chloro-4-fluoro-3-methylbenzoic acid

Specifications

173315-54-3
MFCD11846444
7423351
DBMUFFHLBXKTDM-UHFFFAOYSA-N
2-chloro-4-fluoro-3-methylbenzoic acid
10845315
97%
C8H6ClFO2
1g
benzoic acid, 2-chloro-4-fluoro-3-methyl, 2-chloro-4-fluoro-3-methyl-benzoic acid, 2-chloro-3-methyl-4-fluoro-benzoic acid, 2-chloro-4-fluoro-3-methyl benzoic acid
CC1=C(F)C=CC(C(O)=O)=C1Cl
188.58
188.59
2-Chloro-4-fluoro-3-methylbenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only