missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(dimethylamino)benzaldehyde, ≥95%, Thermo Scientific™

Catalog No. SEW05671DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SEW05671DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW05671DA Supplier Thermo Scientific Chemicals Supplier No. SEW05671DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1424-66-4
Molecular Formula C9H10ClNO
Molecular Weight (g/mol) 183.635
MDL Number MFCD00021051
InChI Key XSQFAWMDRFSIMY-UHFFFAOYSA-N
Synonym 2-chloro-4-dimethylamino benzaldehyde, benzaldehyde, 2-chloro-4-dimethylamino, benzaldehyde,2-chloro-4-dimethylamino, 2-chloro-4-n,n-bis methyl amino benzaldehyde, 4-dimethylamino-2-chlorobenzaldehyde, acmc-209coh, 3-chloro-n,n-dimethyl-4-formylaniline
PubChem CID 74021
IUPAC Name 2-chloro-4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC(=C(C=C1)C=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3N(CH3)C6H3(Cl)CHO
Quantity 1g
Formula Weight 183.64
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(dimethylamino)benzaldehyde
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.