missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-cyanopyridine 98.0+%, TCI America™

Catalog No. C18735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18735G 5 g
C187325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18735G Supplier TCI America Supplier No. C18735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33252-30-1
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.554
MDL Number MFCD00174318
InChI Key QRXBTPFMCTXCRD-UHFFFAOYSA-N
Synonym 2-chloro-4-cyanopyridine, 2-chloroisonicotinonitrile, 2-chloro-4-pyridinecarbonitrile, 2-chloro-isonicotinonitrile, 4-cyano-2-chloropyridine, 4-pyridinecarbonitrile, 2-chloro, 2-chloro-4-cyano pyridine, 2-chlorisonicotinonitril, pubchem2582
PubChem CID 568638
IUPAC Name 2-chloropyridine-4-carbonitrile
SMILES C1=CN=C(C=C1C#N)Cl

Specifications

Melting Point 72°C
Color White-Yellow
Boiling Point 106°C
Quantity 5 g
UN Number 3439
Formula Weight 138.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-cyanopyridine