missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-cyanopyridine, 97%

Catalog No. p-7047862
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1985806 5 g
AAL1985814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1985806 Supplier Thermo Scientific Chemicals Supplier No. L1985806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33252-30-1 | C6H3ClN2 | 138.554 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33252-30-1
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.554
MDL Number MFCD00174318
InChI Key QRXBTPFMCTXCRD-UHFFFAOYSA-N
Synonym 2-chloro-4-cyanopyridine, 2-chloroisonicotinonitrile, 2-chloro-4-pyridinecarbonitrile, 2-chloro-isonicotinonitrile, 4-cyano-2-chloropyridine, 4-pyridinecarbonitrile, 2-chloro, 2-chloro-4-cyano pyridine, 2-chlorisonicotinonitril, pubchem2582
PubChem CID 568638
IUPAC Name 2-chloropyridine-4-carbonitrile
SMILES C1=CN=C(C=C1C#N)Cl

Specifications

Melting Point 69°C to 72°C
Boiling Point 104°C to 106°C (15 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 113927
Formula Weight 138.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-cyanopyridine
Hazard Category H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only