missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-aminopyridine, 97%

Catalog No. p-2698084
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC345450050 5 g Glass bottle
AC345450250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC345450050 Supplier Thermo Scientific Chemicals Supplier No. 345450050
Only null left

CAS: 14432-12-3 | C5H5ClN2 | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14432-12-3
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
InChI Key BLBDTBCGPHPIJK-UHFFFAOYSA-N
Synonym 4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine
PubChem CID 84432
IUPAC Name 2-chloropyridin-4-amine
SMILES C1=CN=C(C=C1N)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 92°C to 94°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 128.56
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-aminopyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breat
EINECSNumber 238-403-

RUO – Research Use Only