missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-diphenylpyrimidine 98.0+%, TCI America™

$78.29 - $164.03

Chemical Identifiers

CAS 2915-16-4
Molecular Formula C16H11ClN2
Molecular Weight (g/mol) 266.728
MDL Number MFCD00030776
InChI Key QNGVEVOZKYHNGL-UHFFFAOYSA-N
PubChem CID 1804739
IUPAC Name 2-chloro-4,6-diphenylpyrimidine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)Cl)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C31231G
View Documents
TCI America
C31231G
1 g N/A
Only null left
 
C31235G
View Documents
TCI America
C31235G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2915-16-4
266.728
QNGVEVOZKYHNGL-UHFFFAOYSA-N
2-chloro-4,6-diphenylpyrimidine
C16H11ClN2
MFCD00030776
1804739
C1=CC=C(C=C1)C2=CC(=NC(=N2)Cl)C3=CC=CC=C3

Specifications

2915-16-4
White-Yellow
MFCD00030776
QNGVEVOZKYHNGL-UHFFFAOYSA-N
2-chloro-4,6-diphenylpyrimidine
1804739
≥98.0% (GC)
2-Chloro-4,6-diphenylpyrimidine
115°C
C16H11ClN2
1 g
C1=CC=C(C=C1)C2=CC(=NC(=N2)Cl)C3=CC=CC=C3
266.728
266.73
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'