missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7125604
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27035G 5 g
C270325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27035G Supplier TCI America Supplier No. C27035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3842-55-5
Molecular Formula C15H10ClN3
Molecular Weight (g/mol) 267.716
MDL Number MFCD05738885
InChI Key DDGPPAMADXTGTN-UHFFFAOYSA-N
Synonym 1,3,5-triazine, 2-chloro-4,6-diphenyl, diphenyl chloro-s-triazine, 4,6-diphenyl-2-chloro-s-triazine, acmc-1ctho, 2-chloro4,6-diphenyl-s-triazine, s-triazine, 2-chloro-4,6-diphenyl, 1,5-triazine, 2-chloro-4,6-diphenyl, 2,6-diphenyl-4-chloro-1,3,5-triazine, 1,3,5-riazine, 2-hloro-4,6-iphenyl, 1,3,5-triazine,2-chloro-4,6-diphenyl
PubChem CID 19698
IUPAC Name 2-chloro-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)C3=CC=CC=C3

Specifications

Melting Point 140°C
Quantity 5 g
Formula Weight 267.72
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,6-diphenyl-1,3,5-triazine