missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethylpyridine, 98+%

Catalog No. AAH6473314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6473314 25 g
AAH6473306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6473314 Supplier Thermo Scientific Chemicals Supplier No. H6473314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 30838-93-8 | C7H8ClN | 141.60 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 30838-93-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD08277279
InChI Key COVGXNSRDOYAJD-UHFFFAOYSA-N
Synonym 2-chloro-4,6-dimethyl pyridine, 2-chloro-4,6-dimethyl-pyridine, pyridine, 2-chloro-4,6-dimethyl, pubchem3919, acmc-1agqf, ksc495g6l, aebsf hydrochloride; pefabloc sc
PubChem CID 12508378
IUPAC Name 2-chloro-4,6-dimethylpyridine
SMILES CC1=CC(C)=NC(Cl)=C1

Specifications

Quantity 25 g
UN Number UN2810
Beilstein 108937
Solubility Information Slightly soluble in water.
Formula Weight 141.6
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-4,6-dimethylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only