Learn More
2-Chloro-4,6-dimethylpyridine-3-carbonitrile, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 14237-71-9 |
|---|---|
| Molecular Formula | C8H7ClN2 |
| Molecular Weight (g/mol) | 166.61 |
| MDL Number | MFCD00051676 |
| InChI Key | RETJKTAVEQPNMH-UHFFFAOYSA-N |
| Synonym | 2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile |
| PubChem CID | 223261 |
| IUPAC Name | 2-chloro-4,6-dimethylpyridine-3-carbonitrile |
| SMILES | CC1=CC(C)=C(C#N)C(Cl)=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC429780050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 14237-71-9 | |
| 166.61 | |
| RETJKTAVEQPNMH-UHFFFAOYSA-N | |
| 223261 | |
| CC1=CC(C)=C(C#N)C(Cl)=N1 |
| C8H7ClN2 | |
| MFCD00051676 | |
| 2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile | |
| 2-chloro-4,6-dimethylpyridine-3-carbonitrile |
Specifications
| 14237-71-9 | |
| 100.0 | |
| C8H7ClN2 | |
| 2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile | |
| CC1=CC(C)=C(C#N)C(Cl)=N1 | |
| 166.61 | |
| 166.61 | |
| 2-chloro-4, 6-dimethylpyridine-3-carbonitrile |
| 96.0 | |
| 96% min. (GC) | |
| MFCD00051676 | |
| RETJKTAVEQPNMH-UHFFFAOYSA-N | |
| 2-chloro-4,6-dimethylpyridine-3-carbonitrile | |
| 223261 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
Harmful in contact with skin.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only