missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4,6-dimethoxy-1,3,5-triazine 97.0+%, TCI America™
$37.29 - $369.08
Chemical Identifiers
| CAS | 3140-73-6 |
|---|---|
| Molecular Formula | C5H6ClN3O2 |
| Molecular Weight (g/mol) | 175.57 |
| MDL Number | MFCD00075607 |
| InChI Key | GPIQOFWTZXXOOV-UHFFFAOYSA-N |
| Synonym | cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy |
| PubChem CID | 18450 |
| IUPAC Name | 2-chloro-4,6-dimethoxy-1,3,5-triazine |
| SMILES | COC1=NC(OC)=NC(Cl)=N1 |
Chemical Identifiers
| 3140-73-6 | |
| 175.57 | |
| GPIQOFWTZXXOOV-UHFFFAOYSA-N | |
| 18450 | |
| COC1=NC(OC)=NC(Cl)=N1 |
| C5H6ClN3O2 | |
| MFCD00075607 | |
| cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy | |
| 2-chloro-4,6-dimethoxy-1,3,5-triazine |
Specifications
| 3140-73-6 | |
| White | |
| C5H6ClN3O2 | |
| 5 g | |
| GPIQOFWTZXXOOV-UHFFFAOYSA-N | |
| 2-chloro-4,6-dimethoxy-1,3,5-triazine | |
| 18450 | |
| ≥97.0% (HPLC,T) | |
| 2-Chloro-4,6-dimethoxy-1,3,5-triazine |
| 78°C | |
| 100°C | |
| MFCD00075607 | |
| cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy | |
| COC1=NC(OC)=NC(Cl)=N1 | |
| 175.57 | |
| 175.57 | |
| Crystalline Powder |
Safety and Handling
TSCA : No