missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4,6-dimethoxy-1,3,5-triazine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 78°C |
| Color | White |
| Boiling Point | 100°C |
| Quantity | 5 g |
| Formula Weight | 175.57 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-4,6-dimethoxy-1,3,5-triazine |
Safety and Handling
| TSCA | No |