missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 74.0°C to 78.0°C |
| Color | White |
| Boiling Point | 99.0°C to 100.0°C (1.0 mmHg) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,sparingly soluble in light petroleum and ethanol |
| Formula Weight | 175.57 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. Causes skin irritation. Causes serious eye damage. Harmful if swallowed. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective |
| EINECSNumber | 221-541- |
RUO – Research Use Only