missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%

Catalog No. AC263000050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC263000050 5 g Glass bottle
AC263000250 25 g Glass bottle
AC263001000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC263000050 Supplier Thermo Scientific Chemicals Supplier No. 263000050

CAS: 3140-73-6 | C5H6ClN3O2 | 175.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3140-73-6
Molecular Formula C5H6ClN3O2
Molecular Weight (g/mol) 175.57
MDL Number MFCD00075607
InChI Key GPIQOFWTZXXOOV-UHFFFAOYSA-N
Synonym cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy
PubChem CID 18450
IUPAC Name 2-chloro-4,6-dimethoxy-1,3,5-triazine
SMILES COC1=NC(OC)=NC(Cl)=N1

Specifications

Melting Point 74.0°C to 78.0°C
Color White
Boiling Point 99.0°C to 100.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,sparingly soluble in light petroleum and ethanol
Formula Weight 175.57
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,6-dimethoxy-1,3,5-triazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye damage.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 221-541-

RUO – Research Use Only