Learn More
2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%
CAS: 3140-73-6 | C5H6ClN3O2 | 175.57 g/mol
$94.48 - $693.11
Chemical Identifiers
| CAS | 3140-73-6 |
|---|---|
| Molecular Formula | C5H6ClN3O2 |
| Molecular Weight (g/mol) | 175.57 |
| MDL Number | MFCD00075607 |
| InChI Key | GPIQOFWTZXXOOV-UHFFFAOYSA-N |
| Synonym | cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy |
| PubChem CID | 18450 |
| IUPAC Name | 2-chloro-4,6-dimethoxy-1,3,5-triazine |
| SMILES | COC1=NC(OC)=NC(Cl)=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2447406
|
Thermo Scientific Chemicals
B2447406 |
5 g |
N/A
|
|
|||||
|
AAB2447414
|
Thermo Scientific Chemicals
B2447414 |
25 g |
N/A
|
|
|||||
|
AAB2447422
|
Thermo Scientific Chemicals
B2447422 |
100 g |
N/A
|
|
|||||
Description
2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.
Solubility
Soluble in water, chloroform and methanol.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 3140-73-6 | |
| 175.57 | |
| GPIQOFWTZXXOOV-UHFFFAOYSA-N | |
| 18450 | |
| COC1=NC(OC)=NC(Cl)=N1 |
| C5H6ClN3O2 | |
| MFCD00075607 | |
| cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy | |
| 2-chloro-4,6-dimethoxy-1,3,5-triazine |
Specifications
| 3140-73-6 | |
| 99°C to 100°C (1 mmHg) | |
| MFCD00075607 | |
| 148988 | |
| Soluble in water,chloroform and methanol. | |
| COC1=NC(OC)=NC(Cl)=N1 | |
| 175.57 | |
| 175.58 | |
| 2-Chloro-4,6-dimethoxy-1,3,5-triazine |
| 75°C to 79°C | |
| C5H6ClN3O2 | |
| 5 g | |
| cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy | |
| GPIQOFWTZXXOOV-UHFFFAOYSA-N | |
| 2-chloro-4,6-dimethoxy-1,3,5-triazine | |
| 18450 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P363-P501c
H302-H317-H318-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 221-541-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only