missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-difluoroaniline 98.0+%, TCI America™

Catalog No. C17771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17771G 1 g
C17775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17771G Supplier TCI America Supplier No. C17771G

Chemical Identifiers

CAS 36556-56-6
Molecular Formula C6H4ClF2N
Molecular Weight (g/mol) 163.552
MDL Number MFCD00142892
InChI Key DUPRZIYJXCCXQZ-UHFFFAOYSA-N
Synonym 6-chloro-2,4-difluoroaniline, 4,6-difluoro-2-chloroaniline, 2-amino-3,5-difluorochlorobenzene, benzenamine, 2-chloro-4,6-difluoro, 2-chloro-4,6-difluorophenylamine, pubchem2940, acmc-1aj0r, ksc226e0f, 2,4-difluoro-6-chloroaniline, 2-chloro-4,6 difluoro aniline
PubChem CID 2734083
IUPAC Name 2-chloro-4,6-difluoroaniline
SMILES C1=C(C=C(C(=C1F)N)Cl)F

Specifications

Color Red-White
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 163.55
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-Chloro-4,6-difluoroaniline