missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4,6-diamino-1,3,5-triazine 95.0+%, TCI America™
Supplier: TCI America C135625G
Specifications
| 2-Chloro-4,6-diamino-1,3,5-triazine | |
| White | |
| MFCD00006054 | |
| 2-chloro-4,6-diamino-1,3,5-triazine, 2,4-diamino-6-chloro-1,3,5-triazine, 2-chloro-4,6-diaminotriazine, 2-chloro-4,6-diamino-s-triazine, 1,3,5-triazine-2,4-diamine, 6-chloro, desethyl-desisopropyl-atrazine, deethyldeisopropylatrazine, deisopropyldeethylatrazine, unii-kvz1hmr84l, 2,4-diamino-6-chloro-s-triazine | |
| NC1=NC(Cl)=NC(N)=N1 | |
| 145.55 | |
| CHEBI:27726 | |
| ≥95.0% (HPLC,N) |
| 3397-62-4 | |
| C3H4ClN5 | |
| 25 g | |
| FVFVNNKYKYZTJU-UHFFFAOYSA-N | |
| 6-chloro-1,3,5-triazine-2,4-diamine | |
| 18831 | |
| 145.55 | |
| Crystalline Powder |
Chemical Identifiers
| 3397-62-4 | |
| 145.55 | |
| FVFVNNKYKYZTJU-UHFFFAOYSA-N | |
| 18831 | |
| 6-chloro-1,3,5-triazine-2,4-diamine |
| C3H4ClN5 | |
| MFCD00006054 | |
| 2-chloro-4,6-diamino-1,3,5-triazine, 2,4-diamino-6-chloro-1,3,5-triazine, 2-chloro-4,6-diaminotriazine, 2-chloro-4,6-diamino-s-triazine, 1,3,5-triazine-2,4-diamine, 6-chloro, desethyl-desisopropyl-atrazine, deethyldeisopropylatrazine, deisopropyldeethylatrazine, unii-kvz1hmr84l, 2,4-diamino-6-chloro-s-triazine | |
| CHEBI:27726 | |
| NC1=NC(Cl)=NC(N)=N1 |
Safety and Handling
TSCA : Yes