missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)quinoxaline, 96%

Catalog No. AAH5054903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5054903 1 g
AAH5054906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5054903 Supplier Thermo Scientific Chemicals Supplier No. H5054903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 254732-51-9 | C9H4ClF3N2 | 232.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 254732-51-9
Molecular Formula C9H4ClF3N2
Molecular Weight (g/mol) 232.59
MDL Number MFCD00243891
InChI Key DSMMAQWRRJQVTQ-UHFFFAOYSA-N
Synonym 2-chloro-3-trifluoromethyl quinoxaline, quinoxaline, 2-chloro-3-trifluoromethyl, chlorotrifluoromethylquinoxaline, 3-chloro-2-trifluoromethyl quinoxaline
PubChem CID 736421
IUPAC Name 2-chloro-3-(trifluoromethyl)quinoxaline
SMILES C1=CC=C2C(=C1)N=C(C(=N2)Cl)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 232.59
Percent Purity 96%
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)quinoxaline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only