missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-quinolinecarboxaldehyde 98.0+%, TCI America™

Catalog No. C22891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22891G 1 g
C22895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22891G Supplier TCI America Supplier No. C22891G

Chemical Identifiers

CAS 73568-25-9
Molecular Formula C10H6ClNO
Molecular Weight (g/mol) 191.614
MDL Number MFCD00130079
InChI Key SDKQWXCBSNMYBN-UHFFFAOYSA-N
Synonym 2-chloro-3-quinolinecarboxaldehyde, 2-chloro-3-quinoline carboxaldehyde, 2-chloro-quinoline-3-carbaldehyde, 2-chloroquinoline-3-carboxaldehyde, 3-quinolinecarboxaldehyde, 2-chloro, 2-chloro-3-quinolinecarbaldehyde, acmc-209or9, 2-chloro-3-formylquinoline, ksc494c9n, 2-chloroquinolin-3-carbaldehyde
PubChem CID 690958
IUPAC Name 2-chloroquinoline-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(C(=N2)Cl)C=O

Specifications

Melting Point 150°C
Quantity 1 g
Formula Weight 191.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-quinolinecarboxaldehyde