missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-pyridinol, 98%, Thermo Scientific™

Chemical Identifiers

CAS 6636-78-8
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD00006235
InChI Key RSOPTYAZDFSMTN-UHFFFAOYSA-N
Synonym 2-chloro-3-hydroxypyridine, 2-chloro-3-pyridinol, 3-pyridinol, 2-chloro, unii-vy212q0s3j, 2-chloro-3-hydroxy pyridine, 2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol, 2-chlorpyridin-3-ol, pubchem2577, 2-chloro-pyridin-3-ol, 2-chloro-pyridine-3-ol
PubChem CID 81136
IUPAC Name 2-chloropyridin-3-ol
SMILES OC1=CC=CN=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC152180250
View Documents Promotion Details
N/A 25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC152181000
View Documents Promotion Details
N/A 100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

6636-78-8
129.54
RSOPTYAZDFSMTN-UHFFFAOYSA-N
81136
OC1=CC=CN=C1Cl
C5H4ClNO
MFCD00006235
2-chloro-3-hydroxypyridine, 2-chloro-3-pyridinol, 3-pyridinol, 2-chloro, unii-vy212q0s3j, 2-chloro-3-hydroxy pyridine, 2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol, 2-chlorpyridin-3-ol, pubchem2577, 2-chloro-pyridin-3-ol, 2-chloro-pyridine-3-ol
2-chloropyridin-3-ol

Specifications

6636-78-8
100.0
97.5% min. (HPLC)
Cl(C5H3N)OH
21, II, 34
RSOPTYAZDFSMTN-UHFFFAOYSA-N
2-chloropyridin-3-ol
81136
98%
97.5
Authentic
C5H4ClNO
MFCD00006235
2-chloro-3-hydroxypyridine, 2-chloro-3-pyridinol, 3-pyridinol, 2-chloro, unii-vy212q0s3j, 2-chloro-3-hydroxy pyridine, 2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol, 2-chlorpyridin-3-ol, pubchem2577, 2-chloro-pyridin-3-ol, 2-chloro-pyridine-3-ol
OC1=CC=CN=C1Cl
129.54
129.55
2-chloro-3-pyridinol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-635-3

SDS
Documents

Documents

RUO – Research Use Only