missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-pyridinecarboxaldehyde, 98%

Catalog No. AC429790050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429790050 5 g Glass bottle
AC429790010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429790050 Supplier Thermo Scientific Chemicals Supplier No. 429790050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36404-88-3 | C6H4ClNO | 141.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36404-88-3
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.56
MDL Number MFCD01315308
InChI Key KHPAGGHFIDLUMB-UHFFFAOYSA-N
Synonym 2-chloro-3-formylpyridine, 2-chloronicotinaldehyde, 2-chloro-3-pyridinecarboxaldehyde, 2-chloro-pyridine-3-carbaldehyde, chloro-2-formyl-3-pyridine, 2-chloropyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-chloro, 2-chloro-3-pyridinecarbaldehyde, 2-chloro-3-pyridinecarboxyaldehyde, 2-chloro-pyridin-3-carbaldehyde
PubChem CID 737607
IUPAC Name 2-chloropyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)Cl)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 47.0°C to 50.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 141.56
Percent Purity 98%
Physical Form Powder or Low Melting Solid
Chemical Name or Material 2-chloro-3-pyridinecarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only